Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Chloroquinoline 98.0+%, TCI America™
SDP

Catalog No. C30855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30855G 5 g
C308525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30855G Supplier TCI America Supplier No. C30855G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 612-61-3
Molecular Formula C9H6ClN
Molecular Weight (g/mol) 163.604
MDL Number MFCD00956370
InChI Key QNGUPQRODVPRDC-UHFFFAOYSA-N
Synonym quinoline, 7-chloro, 7-chloro-quinoline, pubchem19899
PubChem CID 521963
IUPAC Name 7-chloroquinoline
SMILES C1=CC2=C(C=C(C=C2)Cl)N=C1

Specifications

Melting Point 33°C
Color White-Yellow
Boiling Point 268°C
Quantity 5 g
Formula Weight 163.60
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 7-Chloroquinoline
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.