Learn More
Medchemexpress LLC 7-chlorothieno[2,3-c]pyridine | 28948-58-5 | MFCD04973300 | 99.4% | 169.63 g/mol | C7H4ClNS | 5 G
SDP
Catalog No. 5000209926
Encompass_Preferred
7-Chlorothieno[2,3-c]pyridine is a chlorinated thienopyridine derivative used as a synthetic intermediate in pharmaceutical and medicinal chemistry research. It is supplied for research use and commonly used as a building block in organic synthesis.
  • High purity: 99.4% (HPLC).
  • Molecular formula: C7H4ClNS; molecular weight: 169.63 g/mol.
  • Available in multiple package sizes, including 5 G.
  • Intended for use as a synthetic building block in medicinal chemistry research.
Catalog No. 50-002-09926 Supplier Medchemexpress LLC Supplier No. HYW0581825G
7-Chlorothieno[2,3-c]pyridine is a chlorinated thienopyridine derivative used as a synthetic intermediate in pharmaceutical and medicinal chemistry research. It is supplied for research use and commonly used as a building block in organic synthesis.
  • High purity: 99.4% (HPLC).
  • Molecular formula: C7H4ClNS; molecular weight: 169.63 g/mol.
  • Available in multiple package sizes, including 5 G.
  • Intended for use as a synthetic building block in medicinal chemistry research.