Learn More
AdipoGen 7-DCCA
SDP
Catalog No. 502561295
Encompass_Preferred
Chemical. CAS 50995-74-9. Formula C14H15NO4. MW 261.27. Synthetic. 7-Diethylamino-coumarin-3-carboxylic acid is a reagent for the derivatisation of amines and proteins. It has also been used for labelling synthetic peptides for high-throughput detection.
Catalog No. 50-256-1295 Supplier AdipoGen Supplier No. CDXD0036G001
Add to Cart
Add to Cart
Chemical. CAS 50995-74-9. Formula C14H15NO4. MW 261.27. Synthetic. 7-Diethylamino-coumarin-3-carboxylic acid is a reagent for the derivatisation of amines and proteins. It has also been used for labelling synthetic peptides for high-throughput detection.