Learn More
AdipoGen 7-DCCA ethyl ester
SDP
Supplier:  AdipoGen CDXD0126G001
Encompass_Preferred
Chemical. CAS 28705-46-6. Formula C16H19NO4. MW 289.3. Synthetic. Intermediate for the preparation of fluroescent reagents. Laser and scintillation dye. Fluoride chemosensor. Spectral properties lambdaex=418nm lambdaem=461nm, in ethanol. Inhibitor of human monoamine oxidase B hMAO-B.
Catalog No. 50-256-1224