Learn More
Medchemexpress LLC 7-deaza-2'-deoxyadenosine | 60129-59-1 | >=97.0% | 250.25 g/mol | C11H14N4O3 | 50 MG
SDP
Catalog No. 5000210890
Encompass_Preferred
7-deaza-2'-deoxyadenosine is a deoxynucleoside analog used in biochemical research and nucleotide chemistry as a building block for oligonucleotide synthesis and related assays. It is typically supplied as a white solid for laboratory research and is intended for research use only.
  • deoxynucleoside analog for nucleotide chemistry
  • used as a building block in oligonucleotide synthesis
  • supplied as a solid in milligram quantities for laboratory research
  • intended for research use only, not for human or clinical use
Catalog No. 50-002-10890 Supplier Medchemexpress LLC Supplier No. HYW04851350MG
7-deaza-2'-deoxyadenosine is a deoxynucleoside analog used in biochemical research and nucleotide chemistry as a building block for oligonucleotide synthesis and related assays. It is typically supplied as a white solid for laboratory research and is intended for research use only.
  • deoxynucleoside analog for nucleotide chemistry
  • used as a building block in oligonucleotide synthesis
  • supplied as a solid in milligram quantities for laboratory research
  • intended for research use only, not for human or clinical use