Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-deaza-2'-deoxyadenosine | 60129-59-1 | 99.9% | 250.26 g/mol | C11H14N4O3 | 1 ML
SDP

Catalog No. 5000210102
Encompass_Preferred

7-deaza-2'-deoxyadenosine is a deoxynucleoside used as a biochemical reagent and building block for synthesis and oligonucleotide research.

  • Ready-to-use 10 mM solution in DMSO.
  • Supplied as 1 mL vial.
  • High purity, reported as 99.9%.
  • Molecular weight 250.26 g/mol.
  • Molecular formula C11H14N4O3.
  • CAS number 60129-59-1 for unambiguous identification.

Catalog No. 50-002-10102 Supplier Medchemexpress LLC Supplier No. HYW04851310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-deaza-2'-deoxyadenosine is a deoxynucleoside used as a biochemical reagent and building block for synthesis and oligonucleotide research.

  • Ready-to-use 10 mM solution in DMSO.
  • Supplied as 1 mL vial.
  • High purity, reported as 99.9%.
  • Molecular weight 250.26 g/mol.
  • Molecular formula C11H14N4O3.
  • CAS number 60129-59-1 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.