Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-deaza-2'-deoxyadenosine | 60129-59-1 | MFCD00672266 | >97.0% | 250.25 g/mol | C11H14N4O3 | 100 MG
SDP

Catalog No. 5000210885
Encompass_Preferred

7-deaza-2'-deoxyadenosine is a deoxynucleoside analog used as a biochemical reagent for research applications. It is used in studies of nucleic acid structure and function, and has been reported to inhibit DNA synthesis and related polymerase reactions.

  • Oligodeoxynucleotide analog for biochemical research.
  • Reported inhibitor of DNA synthesis and polymerase activity.
  • Molecular formula: C11H14N4O3.
  • Molecular weight: 250.25 g/mol.
  • CAS number: 60129-59-1.
  • Typical purity reported around 98%, minimum 97%.
  • Available as a 100 mg quantity.
  • For research use only; not for diagnostic or therapeutic use.

Catalog No. 50-002-10885 Supplier Medchemexpress LLC Supplier No. HYW048513100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-deaza-2'-deoxyadenosine is a deoxynucleoside analog used as a biochemical reagent for research applications. It is used in studies of nucleic acid structure and function, and has been reported to inhibit DNA synthesis and related polymerase reactions.

  • Oligodeoxynucleotide analog for biochemical research.
  • Reported inhibitor of DNA synthesis and polymerase activity.
  • Molecular formula: C11H14N4O3.
  • Molecular weight: 250.25 g/mol.
  • CAS number: 60129-59-1.
  • Typical purity reported around 98%, minimum 97%.
  • Available as a 100 mg quantity.
  • For research use only; not for diagnostic or therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.