Learn More
Cayman Chemical 7-Deaza-7-iodo-2'-deoxyguanosine, Purity: ≥98%, CAS: 172163-62-1, 1 g
SDP
Catalog No. 502945506
Encompass_Preferred
A nucleoside derivative; has been used as an intermediate in the synthesis of fluorescent nucleotide derivatives for incorporation into single- or double-stranded oligonucleotides
Catalog No. 50-294-5506 Supplier Cayman Chemical Supplier No. 406431 G
A nucleoside derivative; has been used as an intermediate in the synthesis of fluorescent nucleotide derivatives for incorporation into single- or double-stranded oligonucleotides