Learn More
Apexbio Technology LLC 7-Deaza-7-Propargylamino-dATP 50ul (100 mM)
SDP
Catalog No. 5000088038
Encompass_Preferred
7-Deaza-7-Propargylamino-dATP is a chemically modified nucleotide analog derived from deoxyadenosine triphosphate It is designed to enable enzymatic incorporation by DNA polymerases during DNA synthesis thereby facilitating site-specific labeling of DNA 7-Deaza-7-Propargylamino-dATP exerts its biological activity primarily through incorporation into nascent DNA strands introducing a terminal propargyl group that serves as a reactive alkyne handle for conjugation reactions via click chemistry approaches Based on these pharmacological properties 7-Deaza-7-Propargylamino-dATP holds research potential in DNA labeling fluorescent probe generation and studies of DNA polymerase activity specificity and DNA damage detection and repair mechanisms in molecular biology and biomedical research
Catalog No. 50-000-88038 Supplier Apexbio Technology LLC Supplier No. B81210.05
7-Deaza-7-Propargylamino-dATP is a chemically modified nucleotide analog derived from deoxyadenosine triphosphate It is designed to enable enzymatic incorporation by DNA polymerases during DNA synthesis thereby facilitating site-specific labeling of DNA 7-Deaza-7-Propargylamino-dATP exerts its biological activity primarily through incorporation into nascent DNA strands introducing a terminal propargyl group that serves as a reactive alkyne handle for conjugation reactions via click chemistry approaches Based on these pharmacological properties 7-Deaza-7-Propargylamino-dATP holds research potential in DNA labeling fluorescent probe generation and studies of DNA polymerase activity specificity and DNA damage detection and repair mechanisms in molecular biology and biomedical research