Learn More
New England Biolabs, Inc. 7-deaza-dGTP – 1.5 µmol
SDP
Catalog No. 50591164
Encompass
7-deaza-dGTP (7-deaza-2-deoxyguanosine 5-triphosphate) contains a 5 mM solution of 7-deaza-dGTP as a dilithium salt. Nucleotide concentration is determined by measurements of absorbance at 257 nm. Supplied in: Milli-Q water as a lithium salt at (pH 7.0).
Catalog No. 50-591-164 Supplier New England Biolabs, Inc. Supplier No. N0445L
7-deaza-dGTP (7-deaza-2-deoxyguanosine 5-triphosphate) contains a 5 mM solution of 7-deaza-dGTP as a dilithium salt. Nucleotide concentration is determined by measurements of absorbance at 257 nm. Supplied in: Milli-Q water as a lithium salt at (pH 7.0).