Learn More
Apexbio Technology LLC 7-Deaza-dGTP 10ul (5 mM)
SDP
Catalog No. 5000088001
Encompass_Preferred
7-Deaza-dGTP is a deoxyguanosine triphosphate analog characterized by the substitution of the nitrogen atom at position 7 of the purine ring with a carbon atom This compound is designed to reduce the formation of secondary structures in DNA during amplification by weakening hydrogen bonding between bases 7-Deaza-dGTP exerts its biological activity primarily by mitigating secondary-structure-induced inhibition during high GC-content DNA amplification Based on these properties 7-Deaza-dGTP holds research potential in PCR amplification DNA sequencing molecular cloning genomic research and molecular diagnostics
Catalog No. 50-000-88001 Supplier Apexbio Technology LLC Supplier No. B80850.01
7-Deaza-dGTP is a deoxyguanosine triphosphate analog characterized by the substitution of the nitrogen atom at position 7 of the purine ring with a carbon atom This compound is designed to reduce the formation of secondary structures in DNA during amplification by weakening hydrogen bonding between bases 7-Deaza-dGTP exerts its biological activity primarily by mitigating secondary-structure-induced inhibition during high GC-content DNA amplification Based on these properties 7-Deaza-dGTP holds research potential in PCR amplification DNA sequencing molecular cloning genomic research and molecular diagnostics