Learn More
Medchemexpress LLC 7-Deazaguanine | 7355-55-7 | HY-34222 | 99.73% | 150.14 | 25 G
SDP
Catalog No. 5000403001
Encompass_Preferred
7-Deazaguanine (Compound 1) is a highly selective, well-tolerated, brain-penetrant DYRK1A inhibitor. It is promising for research of cancers and Down's syndrome.
  • Highly selective DYRK1A inhibitor
  • Well-tolerated
  • Brain-penetrant
  • Promising for research of cancers
  • Promising for research of Down's syndrome
Catalog No. 50-004-03001 Supplier Medchemexpress LLC Supplier No. 25GHY34222
7-Deazaguanine (Compound 1) is a highly selective, well-tolerated, brain-penetrant DYRK1A inhibitor. It is promising for research of cancers and Down's syndrome.
  • Highly selective DYRK1A inhibitor
  • Well-tolerated
  • Brain-penetrant
  • Promising for research of cancers
  • Promising for research of Down's syndrome