Learn More
Medchemexpress LLC 7-Deazahypoxanthine | 3680-71-5 | 99.9% | C6H5N3O | 25 G
SDP
Catalog No. 5000358644
Encompass_Preferred
7-Deazahypoxanthine is a biochemical reagent designed for life science research. It can be used as a biological material or organic compound, and serves as a pharmaceutical intermediate. Its methylated forms are utilized as regiochemical probes of xanthine oxidase.
  • Biochemical reagent for life science research
  • Suitable as a biological material or organic compound
  • Serves as a pharmaceutical intermediate
  • Methylated forms act as regiochemical probes for xanthine oxidase
Catalog No. 50-003-58644 Supplier Medchemexpress LLC Supplier No. 25GHY77543
7-Deazahypoxanthine is a biochemical reagent designed for life science research. It can be used as a biological material or organic compound, and serves as a pharmaceutical intermediate. Its methylated forms are utilized as regiochemical probes of xanthine oxidase.
  • Biochemical reagent for life science research
  • Suitable as a biological material or organic compound
  • Serves as a pharmaceutical intermediate
  • Methylated forms act as regiochemical probes for xanthine oxidase