Learn More
Medchemexpress LLC 7-Deazaxanthine | 39929-79-8 | 99.7% | C6H5N3O2 | 10 G
SDP
Catalog No. 5000435085
Encompass_Preferred
7-Deazaxanthine (7DX) is an inhibitor of thymidine phosphorylase (TPase). It inhibits the TPase reaction in a concentration-dependent manner with an IC50 value of 40 μM. It also has a significant angiogenesis inhibitory effect.
  • Inhibits thymidine phosphorylase (TPase)
  • Exhibits angiogenesis inhibitory effect
  • High purity of 99.7%
  • Molecular weight of 151.12
Catalog No. 50-004-35085 Supplier Medchemexpress LLC Supplier No. HY7753710G
7-Deazaxanthine (7DX) is an inhibitor of thymidine phosphorylase (TPase). It inhibits the TPase reaction in a concentration-dependent manner with an IC50 value of 40 μM. It also has a significant angiogenesis inhibitory effect.
  • Inhibits thymidine phosphorylase (TPase)
  • Exhibits angiogenesis inhibitory effect
  • High purity of 99.7%
  • Molecular weight of 151.12