Learn More
Medchemexpress LLC HY-113279 100mg Medchemexpress, 7-Dehydrocholesterol CAS:434-16-2 Purity:>98%
SDP
Catalog No. 501969238
Encompass_Preferred
Medchemexpress, HY-113279 100mg 7-Dehydrocholesterol CAS:434-16-2 7-Dehydrocholesterol is biosynthetic precursor of cholesterol and vitamin D3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9238 Supplier Medchemexpress LLC Supplier No. HY113279100MG
Medchemexpress, HY-113279 100mg 7-Dehydrocholesterol CAS:434-16-2 7-Dehydrocholesterol is biosynthetic precursor of cholesterol and vitamin D3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.