Learn More
AdipoGen 7-Dehydrocholesterol
SDP
Supplier:  AdipoGen CDXD0331G010
Encompass_Preferred
Chemical. CAS 434-16-2. Formula C27H44O. MW 384.64. Synthetic Precursor of cholesterol and vitamin D3. It is reduced to cholesterol by the enzyme 3beta-hydroxysterol-Delta7-reductase DHCR7 in the last step of cholesterol biosynthesis. Mutations in the gene encoding DHCR7 lead to increased levels of 7-DHC and the neurodevelopmental syndrome Smith-Lemli-Opitz syndrome SLOS. 7-DHC is used as a biomarker for SLOS screenings. Inhibits human fibroblast sphingomyelinase. Shown to have cytotoxic effects on melanoma cells. In skin, photolysis of 7-DHC by ultraviolet light produces vitamin D3. UV/Vis lambdamax 271, 282, 293nm.
Catalog No. 50-256-1115