Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Diethylamino-3-(4-Maleimidophenyl)-4-Methylcoumarin, Sigma-Aldrich

Catalog No. 01258856 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-856 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-856 Supplier MilliporeSigma Supelco Supplier No. C148425MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CPM is a fluorescent coumarin probe used for thiol monitoring and protein studies.

  • Enables monitoring of thiol release and thiol quantitation.
  • Used for fluorescent detection of coenzyme A and protein thermostability studies.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76877-33-3

Specifications

Concentration ≥95%
Quantity 25 mg
Product Type Fluorescence Reagent
Molecular Weight (g/mol) 402.44 g/mol
Label Type Other Labels or Dyes
Color Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.