Learn More
AdipoGen 7-Diethylamino-3-MD-ethyl-CC
SDP
Catalog No. 502561197
Encompass_Preferred
Chemical. CAS 156571-46-9. Formula C20H21N3O5. MW 383.4. Synthetic. Thiol-reactive fluorescent probe for protein labeling. MDCC has been used for preparing specific probes for inorganic phosphate.
Catalog No. 50-256-1197 Supplier AdipoGen Supplier No. CDXD0198M020
Chemical. CAS 156571-46-9. Formula C20H21N3O5. MW 383.4. Synthetic. Thiol-reactive fluorescent probe for protein labeling. MDCC has been used for preparing specific probes for inorganic phosphate.