Learn More
AdipoGen 7-Diethylamino-3-MD-propyl-CC
SDP
Supplier:  AdipoGen CDXD0199M010
Encompass_Preferred
Chemical. CAS 160291-54-3. Formula C21H23N3O5. MW 397.42. Synthetic. Thiol-reactive probe for protein labelling.
Catalog No. 50-256-1194