Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-(Diethylamino)-3-phenylcoumarin 98.0+%, TCI America™
SDP

Catalog No. D50085G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50085G 5 g
D50081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50085G Supplier TCI America Supplier No. D50085G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 84865-19-0
Molecular Formula C19H19NO2
Molecular Weight (g/mol) 293.366
MDL Number MFCD00408422
InChI Key PSJYODMHCCQXQB-UHFFFAOYSA-N
PubChem CID 688943
IUPAC Name 7-(diethylamino)-3-phenylchromen-2-one
SMILES CCN(CC)C1=CC2=C(C=C1)C=C(C(=O)O2)C3=CC=CC=C3

Specifications

Melting Point 153°C
Quantity 5 g
Formula Weight 293.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-(Diethylamino)-3-phenylcoumarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.