Learn More
Medchemexpress LLC 7- Diethylamino -4- 25mg
SDP
Catalog No. 5000204556
Encompass_Preferred
7-(Diethylamino)-4-(hydroxymethyl)coumarin (compound 10) is an organic compound that can be used as a photosensitizer in organic synthesis[1]
Catalog No. 50-002-04556 Supplier Medchemexpress LLC Supplier No. 25MGHYW094665
7-(Diethylamino)-4-(hydroxymethyl)coumarin (compound 10) is an organic compound that can be used as a photosensitizer in organic synthesis[1]