Learn More
AdipoGen 7-Diethylamino-4-methylcoumari
SDP
Catalog No. 502561225
Encompass_Preferred
Chemical. CAS 91-44-1. Formula C14H17NO2. Molecular Weight 2932.29. 7-Diethylamino-4-methylcoumarin also called Coumarin 1 or Coumarin 460 is a bichromophoric fluorescent laser dye. It can be used as a laser dye, a luminescent phosphor in LEDs, a luminescent probe and sensor in biological applications, and a fluorescer in rewritable optical drives. The efficient fluorescent property of this compound is utilized to sense the Pd2 selectively and sensitively.It is also used as a fluorescent whitening agent FWAs, which is a chemical added to fabrics and paper during manufacture to increase whiteness and brightness. Spectral data lambdaex 373nm ethanol lambdamax 460nm Laser range 450-484nm.
Catalog No. 50-256-1225 Supplier AdipoGen Supplier No. CDXD0124G100
Chemical. CAS 91-44-1. Formula C14H17NO2. Molecular Weight 2932.29. 7-Diethylamino-4-methylcoumarin also called Coumarin 1 or Coumarin 460 is a bichromophoric fluorescent laser dye. It can be used as a laser dye, a luminescent phosphor in LEDs, a luminescent probe and sensor in biological applications, and a fluorescer in rewritable optical drives. The efficient fluorescent property of this compound is utilized to sense the Pd2 selectively and sensitively.It is also used as a fluorescent whitening agent FWAs, which is a chemical added to fabrics and paper during manufacture to increase whiteness and brightness. Spectral data lambdaex 373nm ethanol lambdamax 460nm Laser range 450-484nm.