Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-Diethylamino-4-methylcoumarin | 91-44-1 | 100.0% | 100 G
SDP

Catalog No. 5000410040
Encompass_Preferred

7-Diethylamino-4-methylcoumarin is a laser dye. It is for research use only and not sold to patients.

  • Molecular weight: 231.29
  • Formula: C14H17NO2
  • Appearance: Solid, off-white to light yellow
  • Emission (Em): 470
  • Excitation (Ex): 410

Catalog No. 50-004-10040 Supplier Medchemexpress LLC Supplier No. HYD0082100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Diethylamino-4-methylcoumarin is a laser dye. It is for research use only and not sold to patients.

  • Molecular weight: 231.29
  • Formula: C14H17NO2
  • Appearance: Solid, off-white to light yellow
  • Emission (Em): 470
  • Excitation (Ex): 410

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.