Learn More
Abcam 7-(Diethylamino)coumarin-3-carbonyl azide, 25MG
Catalog No. 502682773
Encompass_Preferred
MW 286.29 Da, Purity >95%. Aldehyde and ketone reactive probe. Suitable for labeling proteins and other carbonyl-containing molecules. Ceramides can be derivatized using 7-(diethylamino)coumarin-3-carbonyl azide with subsequent gradient HPLC separation allowing sensitive optical quantification of individual cellular ceramides. Suitable for use in HPLC labeling of lipids and alkylcycloketones.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-2773 Supplier Abcam Supplier No. AB27554825MG
MW 286.29 Da, Purity >95%. Aldehyde and ketone reactive probe. Suitable for labeling proteins and other carbonyl-containing molecules. Ceramides can be derivatized using 7-(diethylamino)coumarin-3-carbonyl azide with subsequent gradient HPLC separation allowing sensitive optical quantification of individual cellular ceramides. Suitable for use in HPLC labeling of lipids and alkylcycloketones.
The product is subject to the following: Abcam Restricted Use Statement