Learn More
Abcam 7-(Diethylamino)coumarin-3-carbohydrazide, derivatizing agent for carbo x ylic acid detection, 50MG
Catalog No. 502705250
Encompass_Preferred
MW 275.3 g/mol, Purity >95%. Derivatizing agent for detection of carboxylic acid and carbonylated molecules such as lipid peroxidation products (LPP). Has also been used for fluorescence detection of monoterpene alcohols.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5250 Supplier Abcam Supplier No. AB14537750MG
MW 275.3 g/mol, Purity >95%. Derivatizing agent for detection of carboxylic acid and carbonylated molecules such as lipid peroxidation products (LPP). Has also been used for fluorescence detection of monoterpene alcohols.
The product is subject to the following: Abcam Restricted Use Statement