Learn More
Cayman Chemical 7-(Diethylamino)coumarin-3-carboxylic Acid, Purity: ≥95%, CAS: 50995-74-9, 100 mg
SDP
Catalog No. 502913362
Encompass_Preferred
A fluorescent probe; has been used as a FRET acceptor conjugated to neomycin B or tobramycin for use in the analysis of bacterial RNA binding; has been used as a reactive probe for the derivatization of sulfonamides for LIF detection; ex/em = 407/472 nm, respectively
Catalog No. 50-291-3362 Supplier Cayman Chemical Supplier No. 35396100 MG
A fluorescent probe; has been used as a FRET acceptor conjugated to neomycin B or tobramycin for use in the analysis of bacterial RNA binding; has been used as a reactive probe for the derivatization of sulfonamides for LIF detection; ex/em = 407/472 nm, respectively