Learn More
Cayman Chemical 7 DIethylamInocoumarIn3ca 50mg
SDP
Catalog No. 502913569
Encompass_Preferred
A fluorescent probe; has been used as a FRET acceptor conjugated to neomycin B or tobramycin for use in the analysis of bacterial RNA binding; has been used as a reactive probe for the derivatization of sulfonamides for LIF detection; ex/em = 407/472 nm, respectively
Catalog No. 50-291-3569 Supplier Cayman Chemical Supplier No. 3539650 MG
A fluorescent probe; has been used as a FRET acceptor conjugated to neomycin B or tobramycin for use in the analysis of bacterial RNA binding; has been used as a reactive probe for the derivatization of sulfonamides for LIF detection; ex/em = 407/472 nm, respectively