Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-(Dimethylamino)-4-methylcoumarin 98.0+%, TCI America™
SDP

Catalog No. D33555G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33555G 5 g
D335525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33555G Supplier TCI America Supplier No. D33555G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 87-01-4
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.241
InChI Key GZEYLLPOQRZUDF-UHFFFAOYSA-N
PubChem CID 6867
IUPAC Name 7-(dimethylamino)-4-methylchromen-2-one
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)N(C)C

Specifications

Melting Point 142°C
Quantity 5 g
Formula Weight 203.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 7-(Dimethylamino)-4-methylcoumarin
EINECSNumber (5)-3653
RTECSNumber GN6650000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.