Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-dimethylaminocoumarin-4-acetic acid | 80883-54-1 | MFCD00857826 | 98.1% | 247.25 g/mol | C13H13NO4 | 250 MG
SDP

Catalog No. 5000208469
Encompass_Preferred

7-Dimethylaminocoumarin-4-acetic acid is a coumarin-based fluorescent probe used in fluorescence assays, labeling, and assay development. It is supplied as a solid reagent with documented purity, solubility, and storage guidance suitable for in vitro and formulation studies.

  • Fluorescent probe with coumarin core.
  • Purity 98.05%.
  • Molecular weight 247.25 g/mol.
  • Molecular formula C13H13NO4.
  • CAS number 80883-54-1.
  • Recommended solvent: DMSO (100 mg/mL); requires ultrasonic treatment; use newly opened DMSO.
  • Storage: 4°C, protect from light; in solution: -80°C (6 months), -20°C (1 month).
  • Suitable for small-scale fluorescence and labeling experiments.

Catalog No. 50-002-08469 Supplier Medchemexpress LLC Supplier No. 250MGHYW040681
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Dimethylaminocoumarin-4-acetic acid is a coumarin-based fluorescent probe used in fluorescence assays, labeling, and assay development. It is supplied as a solid reagent with documented purity, solubility, and storage guidance suitable for in vitro and formulation studies.

  • Fluorescent probe with coumarin core.
  • Purity 98.05%.
  • Molecular weight 247.25 g/mol.
  • Molecular formula C13H13NO4.
  • CAS number 80883-54-1.
  • Recommended solvent: DMSO (100 mg/mL); requires ultrasonic treatment; use newly opened DMSO.
  • Storage: 4°C, protect from light; in solution: -80°C (6 months), -20°C (1 month).
  • Suitable for small-scale fluorescence and labeling experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.