Learn More
Roboz Surgical #7 DUMONT INOX TIP .17 X .10MM
SDP
Catalog No. NC9174319
Encompass
Dumont Tweezers; Pattern #7; .17 X .10mm Tip Size; 115mm Length; Material: Inox
Catalog No. NC9174319 Supplier Roboz Surgical Supplier No. RS5047
Dumont Tweezers; Pattern #7; .17 X .10mm Tip Size; 115mm Length; Material: Inox