Learn More
TARGETMOL CHEMICALS INC 7-Epi 10-desacetyl paclitaxel
SDP
Catalog No. 502876773
Encompass_Preferred
7-Epi 10-desacetyl paclitaxel (10-Deacetyl-7-epipaclitaxel) shows IC50 values of 0.085 nM, against HeLa cells. Purity 98.05%
Catalog No. 50-287-6773 Supplier TARGETMOL CHEMICALS INC Supplier No. T573810MG
7-Epi 10-desacetyl paclitaxel (10-Deacetyl-7-epipaclitaxel) shows IC50 values of 0.085 nM, against HeLa cells. Purity 98.05%