Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-epi-docetaxel | 153381-68-1 | MFCD15071206 | 98.8% | 807.88 g·mol⁻¹ | C43H53NO14 | 10 MG
SDP

Catalog No. 5000289217
Encompass_Preferred

7-Epi-docetaxel is an epimeric impurity of docetaxel used as a research compound and microtubule/tubulin inhibitor. Supplied as a solid powder in a 10 mg quantity, it is intended for laboratory research use only and not for human or clinical applications.

  • Impurity/epimer of docetaxel used to study tubulin dynamics and cytotoxicity.
  • Solid powder formulation suitable for analytical and cell-based research.
  • CAS number 153381-68-1 for unambiguous identification.
  • Chemical formula C43H53NO14; molecular weight 807.88 g·mol⁻¹.
  • High purity (98.8% reported by supplier) for analytical consistency.
  • Store powder at -20°C for long term; store solutions at -80°C for long term per handling instructions.

Catalog No. 50-002-89217 Supplier Medchemexpress LLC Supplier No. HY1667610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Epi-docetaxel is an epimeric impurity of docetaxel used as a research compound and microtubule/tubulin inhibitor. Supplied as a solid powder in a 10 mg quantity, it is intended for laboratory research use only and not for human or clinical applications.

  • Impurity/epimer of docetaxel used to study tubulin dynamics and cytotoxicity.
  • Solid powder formulation suitable for analytical and cell-based research.
  • CAS number 153381-68-1 for unambiguous identification.
  • Chemical formula C43H53NO14; molecular weight 807.88 g·mol⁻¹.
  • High purity (98.8% reported by supplier) for analytical consistency.
  • Store powder at -20°C for long term; store solutions at -80°C for long term per handling instructions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.