Learn More
Cayman Chemical 7-epI MaresIn 1 100ug
SDP
Catalog No. 502952691
Encompass_Preferred
The inactive 7(S) epimer of Maresin 1; useful as an experimental negative control
Catalog No. 50-295-2691 Supplier Cayman Chemical Supplier No. 13161100 UG
The inactive 7(S) epimer of Maresin 1; useful as an experimental negative control