Learn More
Cayman Chemical 7-epI MaresIn 1 10ug
SDP
Catalog No. 502953552
Encompass_Preferred
The inactive 7(S) epimer of Maresin 1; useful as an experimental negative control
Catalog No. 50-295-3552 Supplier Cayman Chemical Supplier No. 1316110 UG
The inactive 7(S) epimer of Maresin 1; useful as an experimental negative control