Learn More
Cayman Chemical 7-epi Paclitaxel, Purity: ≥98%, CAS: 105454-04-4, 10 mg
SDP
Catalog No. 502981678
Encompass_Preferred
An active metabolite of paclitaxel; exhibits properties comparable to paclitaxel on both cells and in vitro microtubule polymerization
Catalog No. 50-298-1678 Supplier Cayman Chemical Supplier No. 2074110 MG
An active metabolite of paclitaxel; exhibits properties comparable to paclitaxel on both cells and in vitro microtubule polymerization