Learn More
Medchemexpress LLC HY-N0227 10mg Medchemexpress, 7-epi-Taxol CAS:105454-04-4 Purity:>98%
SDP
Catalog No. 501933480
Encompass_Preferred
Medchemexpress, HY-N0227 10mg 7-epi-Taxol CAS:105454-04-4 7-epi-Taxol is an active metabolite of taxol, with activity comparable to that of taxol against cell replication, promoting microtubule bundle formation and against microtubule depolymerization. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3480 Supplier Medchemexpress LLC Supplier No. HYN022710MG
Medchemexpress, HY-N0227 10mg 7-epi-Taxol CAS:105454-04-4 7-epi-Taxol is an active metabolite of taxol, with activity comparable to that of taxol against cell replication, promoting microtubule bundle formation and against microtubule depolymerization. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.