Learn More
AdipoGen 7-Ethoxy-4-CF3-coumarin
SDP
Supplier:  AdipoGen CDXE0003M250
Encompass_Preferred
Chemical. CAS 115453-82-2. Formula C12H9F3O3. MW 258.19. Synthetic. Substrate for monitoring the activity of cytochrome P450 in diverse cells and research issues, including in vitro cytotxity testing, regulation and drug metabolization.
Catalog No. 50-256-1003