Learn More
Cayman Chemical 7-ethoxy-4-Methylcoumarin, Purity: ≥98%, CAS: 87-05-8, 1 g
SDP
Catalog No. 502984043
Encompass_Preferred
A fluorophore used to monitor functional activity of CYP1A1, CYP2B4, and CYP2B6; excitation/emission maxima equals 318 and 377 nm, respectively in methanol and 360 and 449 nm, respectively in dealkylase
Catalog No. 50-298-4043 Supplier Cayman Chemical Supplier No. 195861 G
A fluorophore used to monitor functional activity of CYP1A1, CYP2B4, and CYP2B6; excitation/emission maxima equals 318 and 377 nm, respectively in methanol and 360 and 449 nm, respectively in dealkylase