Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-Ethoxy-4-methylcoumarin | 87-05-8 | 99.4% | 5 G
SDP

Catalog No. 5000410017
Encompass_Preferred

7-Ethoxy-4-methylcoumarin is a coumarin derivative that serves as a substrate probe for mammalian cytochromes P450 1A1, 2B4, and 2B6. It is intended for research use only and not for sale to patients.

  • Functions as a substrate probe for mammalian cytochromes P450 1A1, 2B4, and 2B6.
  • Intended for research use only.
  • Appears as a white to off-white solid.
  • Ships at room temperature.
  • Recommended storage at 4°C, protected from light.
  • Soluble in DMSO at 50 mg/mL.

Catalog No. 50-004-10017 Supplier Medchemexpress LLC Supplier No. HYD00925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Ethoxy-4-methylcoumarin is a coumarin derivative that serves as a substrate probe for mammalian cytochromes P450 1A1, 2B4, and 2B6. It is intended for research use only and not for sale to patients.

  • Functions as a substrate probe for mammalian cytochromes P450 1A1, 2B4, and 2B6.
  • Intended for research use only.
  • Appears as a white to off-white solid.
  • Ships at room temperature.
  • Recommended storage at 4°C, protected from light.
  • Soluble in DMSO at 50 mg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.