Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-Ethoxy-4-methylcoumarin | 87-05-8 | 99.44% | 1 G
SDP

Catalog No. 5000410786
Encompass_Preferred

7-Ethoxy-4-methylcoumarin is a coumarin derivative used as a substrate probe for mammalian cytochromes P450 1A1, 2B4, and 2B6.

  • Coumarin derivative
  • Used as a substrate probe
  • Targets cytochromes P450 1A1, 2B4, and 2B6

Catalog No. 50-004-10786 Supplier Medchemexpress LLC Supplier No. HYD00921G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Ethoxy-4-methylcoumarin is a coumarin derivative used as a substrate probe for mammalian cytochromes P450 1A1, 2B4, and 2B6.

  • Coumarin derivative
  • Used as a substrate probe
  • Targets cytochromes P450 1A1, 2B4, and 2B6

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.