Learn More
Medchemexpress LLC 7-Ethoxy-4-methylcoumarin | 87-05-8 | 99.44% | 100 G
SDP
Catalog No. 5000410006
Encompass_Preferred
7-Ethoxy-4-methylcoumarin is a coumarin derivative that functions as a substrate probe for mammalian cytochromes P450 1A1, 2B4, and 2B6.
  • Coumarin derivative
  • Substrate probe
  • Targets mammalian cytochromes P450 1A1, 2B4, and 2B6
Catalog No. 50-004-10006 Supplier Medchemexpress LLC Supplier No. HYD0092100G
7-Ethoxy-4-methylcoumarin is a coumarin derivative that functions as a substrate probe for mammalian cytochromes P450 1A1, 2B4, and 2B6.
  • Coumarin derivative
  • Substrate probe
  • Targets mammalian cytochromes P450 1A1, 2B4, and 2B6