Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-Ethoxy-4-methylcoumarin | 87-05-8 | 99.44% | 50 G
SDP

Catalog No. 5000410667
Encompass_Preferred

7-Ethoxy-4-methylcoumarin is a coumarin derivative that functions as a substrate probe for mammalian cytochromes P450 1A1, 2B4, and 2B6.

  • Coumarin derivative
  • Substrate probe of mammalian cytochromes P450 1A1, 2B4 and 2B6
  • Molecular weight: 204.22
  • Formula: C12H12O3
  • Appearance: Solid
  • Color: White to off-white
  • SMILES: O=C1C=C(C)C2=CC=C(OCC)C=C2O1

Catalog No. 50-004-10667 Supplier Medchemexpress LLC Supplier No. HYD009250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Ethoxy-4-methylcoumarin is a coumarin derivative that functions as a substrate probe for mammalian cytochromes P450 1A1, 2B4, and 2B6.

  • Coumarin derivative
  • Substrate probe of mammalian cytochromes P450 1A1, 2B4 and 2B6
  • Molecular weight: 204.22
  • Formula: C12H12O3
  • Appearance: Solid
  • Color: White to off-white
  • SMILES: O=C1C=C(C)C2=CC=C(OCC)C=C2O1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.