Learn More
Abcam 7-Etho x y-4-trifluoromethylcoumarin, Fluorescent Cytochrome P450 substrate, 250MG
Catalog No. 502705273
Encompass_Preferred
MW 258.19 g/mol, Purity >98%. Fluorescent substrate for monitoring Cytochrome P450 activity. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5273 Supplier Abcam Supplier No. AB145347250MG
MW 258.19 g/mol, Purity >98%. Fluorescent substrate for monitoring Cytochrome P450 activity. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement