Learn More
Cayman Chemical 7-Ethoxycoumarin, Purity: ≥98%, CAS: 31005-02-4, 1 g
SDP
Catalog No. 502991442
Encompass_Preferred
A CYP450 substrate; undergoes O-deethylation by various CYP isoforms; has been used in the functional characterization of CYPs in cats, rats, and isolated human cornea
Catalog No. 50-299-1442 Supplier Cayman Chemical Supplier No. 312151 G
A CYP450 substrate; undergoes O-deethylation by various CYP isoforms; has been used in the functional characterization of CYPs in cats, rats, and isolated human cornea