Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Ethoxycoumarin, MP Biomedicals™

Catalog No. ICN19517190 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19517190 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19517190 Supplier MP Biomedicals, Inc Supplier No. 0219517190

missing translation for 'validMainframeMsg'

Substrate for fluorometric assay of microsomal monooxygenase activity

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31005-02-4
Molecular Formula C11H10O3
Molecular Weight (g/mol) 190.198
MDL Number MFCD00006877
InChI Key LIFAQMGORKPVDH-UHFFFAOYSA-N
Synonym 7-ethoxycoumarin, 7-ethoxy-2h-chromen-2-one, ethylumbelliferone, 2h-1-benzopyran-2-one, 7-ethoxy, 7-ethoxy-2h-1-benzopyran-2-one, 7-ethoxycoumarine, pubchem8680, coumarin, 7-ethoxy, acmc-209hjx
PubChem CID 35703
ChEBI CHEBI:28184
IUPAC Name 7-ethoxychromen-2-one
SMILES CCOC1=CC2=C(C=C1)C=CC(=O)O2

Specifications

Color White
Quantity 500 mg
Formula Weight 190.2
Physical Form Powder
Chemical Name or Material 7-Ethoxycoumarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.