Learn More
TARGETMOL CHEMICALS INC 7-Ethoxyresorufin 10MG
SDP
Catalog No. 502876000
Encompass_Preferred
Also available in 1 mg, 2 mg, 5 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. 7-Ethoxyresorufin (Resorufin ethyl ether) (7-ER) is a fluorometric substrate and competitive inhibitor of cytochrome P450, particularly CYP1A1. Purity 99.35%
Catalog No. 50-287-6000 Supplier TARGETMOL CHEMICALS INC Supplier No. T2150710MG
Also available in 1 mg, 2 mg, 5 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. 7-Ethoxyresorufin (Resorufin ethyl ether) (7-ER) is a fluorometric substrate and competitive inhibitor of cytochrome P450, particularly CYP1A1. Purity 99.35%