Learn More
Cayman Chemical 7-EthoxyresorufIn 10mg
SDP
Catalog No. 502970825
Encompass_Preferred
A fluorogenic substrate for, and inhibitor of, CYP1A1; upon enzymatic cleavage by CYP1A1, resorufin is released and its fluorescence can be used to quantify CYP1A1 activity; ex/em = 570/580 nm, respectively; a noncompetitive inhibitor of nNOS (Ki = 0.76 µM)
Catalog No. 50-297-0825 Supplier Cayman Chemical Supplier No. 1612210 MG
A fluorogenic substrate for, and inhibitor of, CYP1A1; upon enzymatic cleavage by CYP1A1, resorufin is released and its fluorescence can be used to quantify CYP1A1 activity; ex/em = 570/580 nm, respectively; a noncompetitive inhibitor of nNOS (Ki = 0.76 µM)