Learn More
Enzo Life Sciences 7-Ethoxyresorufin (10mg). CAS: 5725-91-7
SDP
Catalog No. 502010643
Encompass_Preferred
Fluorimetric substrate for cytochrome P450 IA1. Inhibits vasorelaxant responses to acetylcholine. Alternative name: 7-ER. Purity: ≥98%. Solubility: Soluble in DMSO (5mg/ml) or 100% ethanol (5mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0643 Supplier Enzo Life Sciences Supplier No. BMLEI2620010
Fluorimetric substrate for cytochrome P450 IA1. Inhibits vasorelaxant responses to acetylcholine. Alternative name: 7-ER. Purity: ≥98%. Solubility: Soluble in DMSO (5mg/ml) or 100% ethanol (5mg/ml). Long Term Storage: Ambient.