Learn More
Medchemexpress LLC 7-Ethoxyresorufin | 5725-91-7 | 99.6% | 1 ML
SDP
Catalog No. 5000410788
Encompass_Preferred
7-Ethoxyresorufin is a fluorometric substrate and competitive inhibitor of cytochrome P450, especially CYP1A1. It also inhibits NO synthase. This compound is known for its role in biological activity and its fluorescent properties, making it suitable for various research applications.
  • Fluorometric substrate
  • Competitive inhibitor of cytochrome P450
  • Inhibits NO synthase
  • Reduces relaxant actions of acetylcholine
  • Reacts with NADPH to form a fluorescent product
Catalog No. 50-004-10788 Supplier Medchemexpress LLC Supplier No. HYD014510MM/1ML
7-Ethoxyresorufin is a fluorometric substrate and competitive inhibitor of cytochrome P450, especially CYP1A1. It also inhibits NO synthase. This compound is known for its role in biological activity and its fluorescent properties, making it suitable for various research applications.
  • Fluorometric substrate
  • Competitive inhibitor of cytochrome P450
  • Inhibits NO synthase
  • Reduces relaxant actions of acetylcholine
  • Reacts with NADPH to form a fluorescent product