Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-Ethoxyresorufin | 5725-91-7 | 99.6% | 241.24 g/mol | C14H11NO3 | 10 MG
SDP

Catalog No. 5000296779
Encompass_Preferred

7-Ethoxyresorufin is a fluorometric substrate and competitive inhibitor of cytochrome P450 enzymes, especially CYP1A1, and also inhibits nitric oxide synthase; it is used in assays that measure CYP1A activity by conversion to fluorescent resorufin.

  • Fluorometric substrate and competitive inhibitor of CYP1A1.
  • Excitation 335 nm; emission 460 nm, compatible with resorufin detection.
  • High purity (99.61%) suitable for enzymatic and biochemical assays.
  • Soluble in DMF at 2 mg/mL with ultrasonic warming and heating to 60 °C for dissolution.
  • Store at -20 °C and protect from light; in solvent store at -80 °C up to 6 months.

Catalog No. 50-002-96779 Supplier Medchemexpress LLC Supplier No. HYD014510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Ethoxyresorufin is a fluorometric substrate and competitive inhibitor of cytochrome P450 enzymes, especially CYP1A1, and also inhibits nitric oxide synthase; it is used in assays that measure CYP1A activity by conversion to fluorescent resorufin.

  • Fluorometric substrate and competitive inhibitor of CYP1A1.
  • Excitation 335 nm; emission 460 nm, compatible with resorufin detection.
  • High purity (99.61%) suitable for enzymatic and biochemical assays.
  • Soluble in DMF at 2 mg/mL with ultrasonic warming and heating to 60 °C for dissolution.
  • Store at -20 °C and protect from light; in solvent store at -80 °C up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.